missing translation for 'onlineSavingsMsg'
Learn More
Learn More
D4 Dopamine Receptor Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA533483
Code : NEW
Additional Details : Weight : 0.01000kg
Description
Percent identity with other species by BLAST analysis: Human, Gorilla, Gibbon, Monkey, Marmoset (100%) Ferret, Panda (88%) Hamster, Elephant, Horse, Pig (82%).
D4 Dopamine Receptor Polyclonal antibody specifically detects D4 Dopamine Receptor in Human, Non-human Primate samples. It is validated for Immunohistochemistry (Paraffin).Specifications
D4 Dopamine Receptor | |
Polyclonal | |
Unconjugated | |
DRD4 | |
AW125663; D(2C) dopamine receptor; D(4) dopamine receptor; D4DR; D4R; D4RA; Dopamine 4 receptor; Dopamine D4 receptor; dopamine receptor 4; dopamine receptor D4; DRD4; Drd-4; seven transmembrane helix receptor | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
1815 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
1 mg/mL | |
PBS with 0.1% sodium azide | |
P21917 | |
DRD4 | |
Synthetic 17 amino acid peptide from C-terminus of human DRD4 / Dopamine Receptor D4. | |
50 μg | |
Primary | |
Human, Non-human Primate | |
Antibody | |
IgG |