missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dopamine Mouse anti-Chemical, Clone: 2B11, Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA126966
Code : LD
Additional Details : Weight : 0.01000kg
Description
MA1-26966 detects Dopamine. MA1-26966 has been successfully used in inhibition assay procedures. The MA1-26966 immunogen is Dopamine, conjugated to BSA.
Dopamine (C6H3(OH)2-CH2-CH2-NH2) is a catecholamine neurotransmitter in the brain. Its chemical name is 4-(2-aminoethyl)benzene-1,2-diol. Dopamine is a hormone released by the hypothalamus. Its main function is to inhibit the release of prolactin from the anterior lobe of the pituitary. It can be used as a sympathomimetic drug, producing effects such as increased heart rate and blood pressure.Detection of catecholamine-secreting cells.Specifications
Dopamine | |
Monoclonal | |
4.8 mg/mL | |
3,4-dihydroxyphenethylamine; 3-hydroxytyramine; Oxytyramine | |
Dopamine, conjugated to BSA. | |
100 μg | |
Primary | |
Antibody | |
IgG1 |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry | |
2B11 | |
Unconjugated | |
Mouse | |
Protein G | |
RUO | |
4° C, do not freeze | |
Liquid | |
Chemical |