missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dopamine beta Hydroxylase Polyclonal Antibody, Invitrogen™
Sheep Polyclonal Antibody
Brand: Invitrogen PA14655
Code : LD
Additional Details : Weight : 0.01000kg
Description
This antibody is specific for the ∽75 kDa DBH protein in Western blots of human adrenal medulla.
Dopamine beta Hydroxylase Polyclonal antibody specifically detects Dopamine beta Hydroxylase in Human, Non-human Primate samples. It is validated for Western Blot.Specifications
Dopamine beta Hydroxylase | |
Polyclonal | |
0.01M HEPES with 0.15M NaCl, 100μg/mL BSA, 50% glycerol and no preservative; pH 7.5 | |
P09172 | |
DBH | |
Synthetic peptide corresponding to the N-terminal region of human DBH conjugated to KLH | |
100 μL | |
Primary | |
Human, Non-human Primate | |
Antibody | |
IgG |
Western Blot | |
Unconjugated | |
DBH | |
Dbh; DBM; dopamine beta hydroxylase; Dopamine beta hydroxylase (dopamine beta-monooxygenase); dopamine beta-hydroxylase; dopamine beta-hydroxylase (dopamine beta-monooxygenase); dopamine beta-hydroxylase precursor; dopamine beta-hydroxylase precursor (EC 1.14.17.1); dopamine beta-monooxygenase; dopamine beta-monooxygenase precursor (EC 1.14.17.1); Dopamine-Beta-hydroxylase; DOPBHY; mixed function oxidase; Soluble dopamine beta-hydroxylase | |
Sheep | |
Affinity chromatography | |
RUO | |
1621 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |