missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FILIP1L Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA532021
Code : PN
Additional Details : Weight : 0.01000kg
Description
Recommended positive controls: HCT116, MCF-7. Predicted reactivity: Mouse (89%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
FILIP1L Polyclonal antibody specifically detects FILIP1L in Human samples. It is validated for Western Blot.Specifications
FILIP1L | |
Polyclonal | |
Unconjugated | |
FILIP1L | |
130 kDa GPBP-interacting protein; 4631422O05Rik; 90 kDa GPBP-interacting protein; COL4A3BPIP; DOC1; DOC-1; downregulated in ovarian cancer 1-like; filamin A interacting protein 1 like; filamin A interacting protein 1-like; filamin A-interacting protein 1-like; FILIP1L; GIP130; GIP130a; GIP130b; GIP130c; GIP90; Protein down-regulated in ovarian cancer 1; protein down-regulated in ovarian cancer 1 homolog; RGD1565927 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
11259 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot | |
1 mg/mL | |
PBS with 1% BSA, 20% glycerol and 0.01% thimerosal; pH 7 | |
Q4L180 | |
FILIP1L | |
Recombinant fragment corresponding to a region within amino acids 252 and 487 of Human DOC1 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |