Learn More
DLL1 Mouse anti-Rat, Clone: 409005, R&D Systems™
Mouse Monoclonal Antibody
Brand:  RD Systems (Bio-Techne) MAB3970
Code : Z1
Additional Details : Weight : 0.00010kg
 View more versions of this product
Product Code. 13440346
  • £249.49 / 100µg
Estimated Shipment
28-06-2019
Description
Description
DLL1 Monoclonal antibody MAB3970 Specifically detects DLL1 in Rat samples. It is validated for Western Blotting
Specifications
Specifications
DLL1
409005
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Purified
DLL1
Mouse myeloma cell line NS0-derived recombinant rat DLL1 Gln18-Trp537 Accession # P97677
Protein A or G purified from hybridoma culture supernatant
RUO
Primary
28514
Reconstitute at 0.5 mg/mL in sterile PBS.
Western Blot
LYOPH
Western Blot 1 ug/mL
P97677
Delta, delta (Drosophila)-like 1, Delta 1, Delta1, delta-like 1 (Drosophila), delta-like protein 1, DL1, Drosophila Delta homolog 1, H-Delta-1
Mouse
IgG1
100 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Monoclonal
Detects rat DLL1 in direct ELISAs and Western blots. In direct ELISAs and Western blots, no cross-reactivity with recombinant rat (rr) Jagged-1, recombinant mouse (rm) Jagged-2, rmNotch-3, rrNotch-2, and rrNotch-1 is observed.
Rat