missing translation for 'onlineSavingsMsg'
Learn More

Dkk-4 Mouse anti-Human, Clone: 185709, R&D Systems™

Mouse Monoclonal Antibody

Brand:  RD Systems (Bio-Techne) MAB1269-500

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13488505

  • £353.77 / 500µg


Estimated Shipment
04-10-2018
Add to basket

Description

Description

Dkk-4 Monoclonal antibody MAB1269500 Specifically detects Dkk-4 in Human samples. It is validated for ELISA Capture, Western Blotting
Specifications

Specifications

Dkk-4
185709
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Purified
DKK4
Mouse myeloma cell line NS0-derived recombinant human Dkk-4, Leu19-Leu224, Accession # Q9UBT3
Protein A or G purified from hybridoma culture supernatant
RUO
Primary
27121
Reconstitute at 0.5 mg/mL in sterile PBS.
Western Blot, ELISA
LYOPH
Western Blot 1 ug/mL, ELISA Capture (Matched Antibody Pair) 2-8 ug/mL
Q9UBT3
dickkopf (Xenopus laevis) homolog 4, dickkopf homolog 4 (Xenopus laevis), dickkopf-4, dickkopf-related protein 4, Dkk4, DKK-4, hDkk-4, MGC129562, MGC129563
Mouse
IgG2b
500 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20 to -70 °C under sterile conditions after reconstitution.
Monoclonal
Detects human Dkk-4 in ELISAs and Western blots. In direct ELISAs and Western blots, less than 2% cross-reactivity with recombinant mouse (rm) Dkk-4 is observed and no cross-reactivity with recombinant human (rh) Dkk-1 or rhDkk-3 is observed. In sandwich immunoassays, less than 2% cross-reactivity with rmDkk-4 is observed and no cross-reactivity with rhDkk-1, recombinant rat Dkk-1, rmDkk-1, or rhDkk-3 is observed.
Human
Videos