Learn More
Dkk-3 Mouse anti-Human, Clone: 138726, R&D Systems™
Mouse Monoclonal Antibody
Brand:  RD Systems (Bio-Techne) MAB1118-500
Code : Z1
Additional Details : Weight : 0.00010kg
 View more versions of this product
Product Code. 13488355
  • £363.01 / 500µg
Estimated Shipment
03-10-2018
Add to basket
Description
Description
Dkk-3 Monoclonal antibody MAB1118500 Specifically detects Dkk-3 in Human samples. It is validated for Western Blotting
Specifications
Specifications
Dkk-3
Unconjugated
Murine
IgG1
500μg
Primary
Human
138726
Purified
Mouse myeloma cell line NS0-derived recombinant human Dkk-3. Ala22-Ile350
Protein A or G purified from hybridoma culture supernatant