Learn More
DISP1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA543505
Code : NEW
Additional Details : Weight : 0.00010kg
Description
Peptide sequence: FVLCDVWNYT KFDKPHAETS ETVSITLQHA ALSMFVTSFT TAAAFYANYV Sequence homology: Horse: 93%; Human: 100%; Mouse: 93%; Pig: 86%; Rabbit: 100%; Rat: 86%; Zebrafish: 79%
DISP1 Polyclonal antibody specifically detects DISP1 in Human samples. It is validated for Western Blot.Specifications
DISP1 | |
Polyclonal | |
Unconjugated | |
DISP1 | |
1190008H24Rik; chameleon; con; Disp; Disp1; Dispa; dispatched 1; dispatched A; dispatched homolog 1; dispatched homolog 1 (Drosophila); dispatched RND transporter family member 1; DKFZP434I0428; FLJ43740; helix; hlx; hypothetical protein LOC619201; Icb; Icbins; Mdispa; MGC104180; MGC13130; MGC16796; multipass transmembrane protein; Disp1; protein chameleon; Protein dispatched homolog 1; protein dispatched homolog 1 pseudogene; twelve-pass transmembrane protein; zgc:111866 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
84976 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q96F81 | |
DISP1 | |
Synthetic peptide directed towards the middle region of human DISP1 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |