Learn More
DHX37 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA541296
Code : NEW
Additional Details : Weight : 0.00010kg
Description
Peptide sequence: PLTKKEKKVL QKILEQKEKK SQRAEMLQKL SEVQASEAEM RLFYTTSKLG Sequence homology: Dog: 93%; Horse: 93%; Human: 100%; Mouse: 93%; Pig: 79%; Rabbit: 79%; Rat: 93%
DHX37 Polyclonal antibody specifically detects DHX37 in Human samples. It is validated for Western Blot.Specifications
DHX37 | |
Polyclonal | |
Unconjugated | |
DHX37 | |
DDX37; DEAD/DEAH box helicase DDX37; DEAD/H (Asp-Glu-Ala-Asp/His) box polypeptide 37; DEAH (Asp-Glu-Ala-His) box polypeptide 37; DEAH box protein 37; DEAH-box helicase 37; DHX37; FLJ41974; Gm1050; Gm451; KIAA1517; LOW QUALITY PROTEIN: probable ATP-dependent RNA helicase DHX37; MGC2695; MGC4322; MGC46245; mKIAA1517; Probable ATP-dependent RNA helicase DHX37 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
57647 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q8IY37 | |
DHX37 | |
Synthetic peptide directed towards the N-terminal of human DHX37 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |