Learn More
Desmoglein Mouse, Unlabeled, Clone: 62, BD
Mouse Monoclonal Antibody
Brand: BD Biosciences 610273
Additional Details : Weight : 0.01000kg
Description
Desmoglein (Dsg1) is a transmembrane glycoprotein found in desmosome junctions. Desmosomes and adherens junctions are plasma membrane specializations involved in cell-cell adhesion. They serve as membrane anchorage domains for intermediate filaments and microfilaments, respectively. Actin microfilaments anchor at adherens junctions, which occur in many sizes, shapes, and positions. These junctions generally contain E- or N-Cadherin whose cytoplasmic region is associated with cytoplasmic proteins such as α- and β- catenin, vinculin, α-actinin, and radixin. Intermediate filaments (IFs) are anchored at desmosomes which contain desmosome-specific cadherin family members, known as desmocollin (Dsc) and desmoglein. These proteins associate with cytoplasmic proteins such as desmoplakin and plakoglobin (γ-Catenin). Plakoglobin is the only junctional protein found at both desmosomes and adherens junctions and, therefore, plays a crucial role in cell adhesion.
Immunofluorescence, Immunohistochemistry, Immunoprecipitation, Western Blotting
Specifications
Desmoglein | |
Monoclonal | |
250μg/mL | |
Dsg1 | |
Mouse | |
Affinity Purified | |
RUO | |
Primary | |
Store undiluted at -20°C. |
Western Blot | |
62 | |
Unconjugated | |
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide. | |
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide. | |
50 μg | |
Cell Biology | |
Canine, Chicken, Drosophila, Human, Mouse, Rat | |
IgG2b |
For Research Use Only.