Learn More
Desmin Monoclonal Antibody (D33), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA513259
Code : LE
Additional Details : Weight : 0.01000kg
Description
MA5-13259 targets Desmin in IF and IHC (P) applications and shows reactivity with Bovine, Canine, Chicken, Feline, Hamster, Human, Non-human primate, and Rat samples. The MA5-13259 immunogen is desmin from human muscle.
Desmin Monoclonal antibody specifically detects Desmin in Bovine, Canine, Chicken, Feline, Hamster, Human, Non-human Primate, Rat samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot.Specifications
Desmin | |
Monoclonal | |
Conc. Not Determined | |
tissue culture supernatant with 0.09% sodium azide | |
O62654, P02542, P17661, P48675, Q5XFN2 | |
DES | |
Desmin from human muscle | |
RUO | |
100764990, 101097010, 1674, 280765, 395906, 497091, 64362 | |
4° C | |
Liquid |
Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot | |
D33 | |
Unconjugated | |
DES | |
CMD1I; CSM1; CSM2; DES; Desmin; desmin-like protein; FLJ12025; FLJ39719; FLJ41013; FLJ41793; I79_018932; intermediate filament protein; LGMD2R; MFM1; muscle-specific intermediate filament desmin; muscle-specific intermediate filament protein; mutant desmin p.K241E; SCPNK; similar to desmin | |
Mouse | |
500 μL | |
Primary | |
Bovine, Canine, Chicken, Feline, Hamster, Human, Non-human Primate, Rat | |
Antibody | |
IgG1 κ |