Learn More
DEP-1/CD148 Mouse anti-Human, Clone: 261922, R&D Systems™
Mouse Monoclonal Antibody
Brand:  RD Systems (Bio-Techne) MAB19341
Code : Z1
Additional Details : Weight : 0.00010kg
 View more versions of this product
Product Code. 13439045
  • £288.60 / 100µg
Estimated Shipment
14-09-2020
Description
Description
DEP-1/CD148 Monoclonal antibody MAB19341 Specifically detects DEP-1/CD148 in Human samples. It is validated for Flow Cytometry
Specifications
Specifications
DEP-1/CD148
261922
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Purified
PTPRJ
E. coli-derived recombinant human DEP-1/CD148 Arg997-Ala1337 Accession # Q12913
Protein A or G purified from hybridoma culture supernatant
RUO
Primary
5795
Reconstitute at 0.5 mg/mL in sterile PBS.
Flow Cytometry, CyTOF
LYOPH
Flow Cytometry 0.25 ug/10^6 cells, CyTOF-ready
Q12913
CD148, CD148 antigen, Density-enhanced phosphatase 1, DEP1, DEP-1, DEP1EC 3.1.3.48, HPTP eta, HPTPeta, HPTP-eta, human density enhanced phosphatase-1, protein tyrosine phosphatase, receptor type, J, protein tyrosine phosphatase, receptor type, J polypeptide, Protein-tyrosine phosphatase eta, Protein-tyrosine phosphatase receptor type J, PTPRJ, receptor-type tyrosine-protein phosphatase eta, R-PTP-eta, R-PTP-J, SCC1, susceptibility to colon cancer 1, mouse, homolog of
Mouse
IgG2b
100 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Monoclonal
Detects human DEP-1/CD148.