missing translation for 'onlineSavingsMsg'
Learn More

Dectin-2/CLEC6A α Isoform Rat anti-Mouse, Clone: 217611, R&D Systems™

Rat Monoclonal Antibody

Brand:  RD Systems (Bio-Techne) MAB1525

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13458705

  • £363.01 / 500µg


Estimated Shipment
03-12-2018
Add to basket

Description

Description

Dectin-2/CLEC6A α Isoform Monoclonal antibody MAB1525 Specifically detects Dectin-2/CLEC6A α Isoform in Mouse samples. It is validated for Western Blotting
Specifications

Specifications

Dectin-2/CLEC6A
217611
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Purified
Clec4n
Mouse myeloma cell line NS0-derived recombinant mouse Dectin-2/CLEC6A, Phe43-Leu209, Accession # Q9JKF4
Protein A or G purified from hybridoma culture supernatant
RUO
Primary
93978
Reconstitute at 0.5 mg/mL in sterile PBS.
Western Blot
LYOPH
Western Blot 1 ug/mL
Q9JKF4
CLEC4N, CLEC6A, CLECSF10, C-type (calcium dependent, carbohydrate-recognition domain) lectin, superfamilymember 10, C-type lectin domain family 6 member A, C-type lectin domain family 6, member A, C-type lectin superfamily member 10, DC-associated C-type lectin 2, dectin 2, Dectin-2, DECTIN2, Dendritic cell-associated C-type lectin 2
Rat
IgG1
500 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20 to -70 °C under sterile conditions after reconstitution.
Monoclonal
Detects mouse Dectin-2/CLEC6A in direct ELISAs and Western blots. In direct ELISAs and Western blots, no cross-reactivity with recombinant human (rh) Dectin-1 or rhDLEC is observed.
Mouse
Videos

For Research Use Only