missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Decorin Polyclonal Antibody, Invitrogen™
Goat Polyclonal Antibody
Brand: Invitrogen PA519151
Code : A0
Additional Details : Weight : 0.01000kg
Description
This antibody is predicted to react with bovine, porcine and Cavia rodent based on sequence homology.
Decorin Polyclonal antibody specifically detects Decorin in Human samples. It is validated for Immunohistochemistry (Paraffin), Western Blot.Specifications
Decorin | |
Polyclonal | |
Unconjugated | |
DCN | |
Bone proteoglycan II; CSCD; DC; Dcn; DCN protein; DCN1C; Decorin; decorin isoform a preproprotein; Dermatan sulfate proteoglycan-II; dermatan sulfate proteoglycan-II (decorin); dermatan sulphate proteoglycans II; Dermatan sulphate proteoglycans II (DSPG2); DSPG; DSPG2; mDcn; PG40; PGII; PGS2; PG-S2; proteoglycan core protein; proteoglycan core protein; binds type I collagen fibers and TGF-beta; decoron; similar to bone proteoglycan II; similar to decorin; SLRR1B; Small leucine rich protein 1B; small leucine-rich protein 1B; unnamed protein product | |
Goat | |
Antigen affinity chromatography | |
RUO | |
1634 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.5 mg/mL | |
TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
P07585 | |
DCN | |
Synthetic peptide sequence (KISRVDAASLKGLNN) corresponding to the internal amino acids of DCN | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG |