Learn More
Decorin Rat anti-Mouse, Clone: 161026, R&D Systems™
Rat Monoclonal Antibody
Brand:  RD Systems (Bio-Techne) MAB1060
Code : Z1
Additional Details : Weight : 0.00010kg
 View more versions of this product
Product Code. 13438295
  • £337.93 / 500µg
Estimated Shipment
08-07-2019
Description
Description
Decorin Monoclonal antibody MAB1060 Specifically detects Decorin in Mouse samples. It is validated for Western Blotting
Specifications
Specifications
Decorin
161026
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Purified
DCN
Mouse myeloma cell line NS0-derived recombinant mouse Decorin Gly17-Lys354 Accession # P28654
Protein A or G purified from hybridoma culture supernatant
RUO
Primary
1634
Reconstitute at 0.5 mg/mL in sterile PBS.
Western Blot
LYOPH
Western Blot 1 ug/mL
P28654
Bone proteoglycan II, CSCD, DCN, decorin, decorin proteoglycan, dermatan sulphate proteoglycans II, DSPG2, PG40, PGII, PG-II, PGS2, PG-S2, proteoglycan core protein, SLRR1B, SLRR1BPGS2, small leucine-rich protein 1B
Rat
IgG2a
500 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Monoclonal
Detects mouse Decorin in direct ELISAs and Western blots. In Western blots, this antibody does not cross-react with recombinant human Decorin.