Learn More
Decorin Mouse anti-Human, Clone: 115424, R&D Systems™
Mouse Monoclonal Antibody
Brand:  RD Systems (Bio-Techne) MAB1432
Code : Z1
Additional Details : Weight : 0.00010kg
 View more versions of this product
Product Code. 13448635
  • £376.32 / 500µg
Estimated Shipment date
from Supplier 21-11-2019
Description
Description
Decorin Monoclonal antibody MAB1432 Specifically detects Decorin in Human samples. It is validated for ELISA Capture
Specifications
Specifications
Decorin
115424
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Purified
DCN
S. frugiperda insect ovarian cell line Sf 21-derived recombinant human Decorin Gly17-Lys359 Accession # P07585
Protein A or G purified from hybridoma culture supernatant
RUO
Primary
1634
Reconstitute at 0.5 mg/mL in sterile PBS.
ELISA
LYOPH
ELISA Capture (Matched Antibody Pair) 2-8 ug/mL
P07585
Bone proteoglycan II, CSCD, DCN, decorin, decorin proteoglycan, dermatan sulphate proteoglycans II, DSPG2, PG40, PGII, PG-II, PGS2, PG-S2, proteoglycan core protein, SLRR1B, SLRR1BPGS2, small leucine-rich protein 1B
Mouse
IgG1
500 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Monoclonal
Detects human Decorin in ELISAs.