missing translation for 'onlineSavingsMsg'
Learn More

DDR1 Mouse anti-Human, Clone: 290420, R&D Systems™

Mouse Monoclonal Antibody

Brand:  RD Systems (Bio-Techne) MAB2396

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13459405

  • £264.01 / 100µg


Estimated Shipment
29-11-2018
Add to basket

Description

Description

DDR1 Monoclonal antibody MAB2396 Specifically detects DDR1 in Human samples. It is validated for Immunoprecipitation
Specifications

Specifications

DDR1
290420
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Purified
DDR1
Mouse myeloma cell line NS0-derived recombinant human DDR1, Asp19-Thr416, Accession # Q08345
Protein A or G purified from hybridoma culture supernatant
RUO
Primary
780
Reconstitute at 0.5 mg/mL in sterile PBS.
Immunoprecipitation
LYOPH
Immunoprecipitation 25 ug/mL
Q08345
CAK, CD167 antigen-like family member A, CD167a, CD167a antigen, DDR, discoidin domain receptor family, member 1, discoidin domain receptor tyrosine kinase 1, Discoidin receptor tyrosine kinase, EC 2.7.10, EC 2.7.10.1, EDDR1, ENTRK4, Epithelial discoidin domain receptor 1, HGK2, MCK10, MCK-10, NTRK4, Protein-tyrosine kinase 3A, PTK3A, PTK3A protein tyrosine kinase 3A, receptor, type 4, RTK6, TRK E, TrkE
Mouse
IgG2a
100 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20 to -70 °C under sterile conditions after reconstitution.
Monoclonal
Detects human DDR1 in direct ELISAs and Western blots. In direct ELISAs and Western blots, this antibody does not cross-react with recombinant human DDR2.
Human
Videos

For Research Use Only