Learn More
DcTRAIL R1/TNFRSF23 Rat anti-Mouse, Clone: 270028, R&D Systems™
Rat Monoclonal Antibody
Brand:  RD Systems (Bio-Techne) MAB23781
Code : Z1
Additional Details : Weight : 0.00010kg
 View more versions of this product
Product Code. 13449395
  • £277.83 / 100µg
Estimated Shipment date
from Supplier 20-11-2019
Description
Description
DcTRAILR1/TNFRSF23 Monoclonal antibody specifically detects DcTRAILR1/TNFRSF23 in Mouse samples. It is validated for Western Blot.
Specifications
Specifications
DcTRAILR1/TNFRSF23
270028
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Purified
Rat
IgG2a
100 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Monoclonal
Detects mouse DcTRAIL R1/TNFRSF23 in direct ELISAs and Western blots. In Western blots, no cross-reactivity with recombinant human (rh) BAFF R, recombinant mouse (rm) BAFF R, rhCD27, rmCD27, rhCD30, rmCD30, rhCD40, rmCD40, rhDR3, rmDR3, rhDR6, rhEDAR, rmEDAR, rhFas, rmFas, recombinant rat Fas, recombinant feline Fas, rhGITR, rmGITR, rhLymphotoxin beta R, rhNGF R, rmNGF R, rhRELT, or rhTROY is observed.
Mouse
Western Blot
LYOPH
Western Blot 1 ug/mL
Q9ER63
Tnfrsf23
Mouse myeloma cell line NS0-derived recombinant mouse DcTRAIL R1/TNFRSF23 Met30-Ser156 Accession # Q9ER63
Protein A or G purified from hybridoma culture supernatant
RUO
Primary
8718
Reconstitute at 0.5 mg/mL in sterile PBS.