Learn More
DcR3/TNFRSF6B Mouse anti-Human, Clone: 229817, R&D Systems™
Mouse Monoclonal Antibody
Brand:  RD Systems (Bio-Techne) MAB21351
Code : Z1
Additional Details : Weight : 0.00010kg
 View more versions of this product
Product Code. 13439205
  • £390.72 / 500µg
Estimated Shipment
19-08-2020
Description
Description
DcR3/TNFRSF6B Monoclonal antibody MAB21351 Specifically detects DcR3/TNFRSF6B in Human samples. It is validated for ELISA Capture, Western Blotting
Specifications
Specifications
DcR3/TNFRSF6B
229817
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Purified
TNFRSF6B
Mouse myeloma cell line NS0-derived recombinant human DcR3/TNFRSF6B Val24-His300 Accession # O95407
Protein A or G purified from hybridoma culture supernatant
RUO
Primary
8771
Reconstitute at 0.5 mg/mL in sterile PBS.
Western Blot, ELISA
LYOPH
Western Blot 1 ug/mL, ELISA Capture (Matched Antibody Pair) 2-8 ug/mL
O95407
DCR3Decoy receptor for Fas ligand, Decoy receptor 3, M68, M68DJ583P15.1.1, TNFRSF6B, TR6, TR6DcR3, tumor necrosis factor receptor superfamily member 6B, tumor necrosis factor receptor superfamily, member 6b, decoy
Mouse
IgG1
500 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Monoclonal
Detects human DcR3/TNFRSF6B in ELISAs and Western blots. In Western blots, no cross-reactivity with recombinant human (rh) OPG, recombinant mouse (rm) OX40, rhRANK, rhRELT, rhTAJ, rhTNF RI, rhTNF RII, rhTRADD, rhTWEAK R, rhTRAIL RI, rhTRAIL R2, rhTRAIL R3, or rhTRAIL R4 is observed.