missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytomegalovirus pp65 Monoclonal Antibody (1-L-11), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA17597
Code : LD
Additional Details : Weight : 0.01000kg
Description
MA1-7597 detects HCMV from virus samples. MA1-7597 has been successfully used in ELISA, Western blot procedures. The MA1-7597 immunogen is HCMV infected cells. Store short-term at 4oC, long-term at -80oC.
Cytomegalovirus pp65 Monoclonal antibody specifically detects Cytomegalovirus pp65 in Virus samples. It is validated for ELISA, Immunocytochemistry, Western Blot.Specifications
Cytomegalovirus pp65 | |
Monoclonal | |
1 mg/mL | |
PBS with 0.01% sodium azide; pH 7.2 | |
Mouse | |
Protein A | |
RUO | |
Virus | |
Antibody | |
IgG1 |
ELISA, Immunocytochemistry, Western Blot | |
1-L-11 | |
Unconjugated | |
CMV; Cytomegalovirus | |
HCMV infected cells. | |
100 μg | |
Primary | |
4°C short term, -80°C long term | |
Liquid |