missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytokeratin 4 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA529463
Code : PN
Additional Details : Weight : 0.01000kg
Description
Recommended positive controls: HeLa. Predicted reactivity: Dog (88%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Cytokeratin 4 Polyclonal antibody specifically detects Cytokeratin 4 in Human samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot.Specifications
Cytokeratin 4 | |
Polyclonal | |
Unconjugated | |
KRT4 | |
57kd keratin; 57kDa keratin; AW108092; cb59; CK4; CK-4; CYK4; cytokeratin; cytokeratin 4; cytokeratin-4; Cytoskeletal 57 kDa keratin; etID20321.6; intermediate filament protein; K4; Kb4; keratin 4; keratin 4, type II; keratin 8; keratin complex 2, basic, gene 4; keratin, type II cytoskeletal 4; Keratin-4; Krt-2.4; Krt2-4; Krt4; krt4 {ECO:0000250; krt4 protein; krt8; type II basic cytokeratin; type II keratin Kb4; type-II keratin Kb4; UniProtKB:P19013}; WSN1; wu:fa91a11; wu:fd19e09; wu:fl23c09; zf-K8 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
3851 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot | |
1 mg/mL | |
0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7 | |
P19013 | |
KRT4 | |
Recombinant protein fragment corresponding to a region within amino acids 317 and 505 of Human Cytokeratin 4 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |