Learn More
Cytokeratin 19 Monoclonal Antibody (4F12G9), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA515477
Code : A0
Additional Details : Weight : 0.01000kg
Description
MA5-15477 targets Cytokeratin 19 in indirect ELISA, WB applications and shows reactivity with Human samples. The MA5-15477 immunogen is purified recombinant fragment of Cytokeratin 19 (aa80-400) expressed in E. Coli strain. <br>. MA5-15477 detects Cytokeratin 19 which has a predicted molecular weight of approximately 44kDa.
Cytokeratin 19 Monoclonal antibody specifically detects Cytokeratin 19 in Human samples. It is validated for ELISA, Immunocytochemistry, Western Blot.Specifications
Cytokeratin 19 | |
Monoclonal | |
Conc. Not Determined | |
ascites with 0.03% sodium azide | |
P08727 | |
KRT19 | |
Purified recombinant fragment of Cytokeratin 19 (aa80-400) expressed in E. Coli strain. | |
RUO | |
3880 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
ELISA, Immunocytochemistry, Western Blot | |
4F12G9 | |
Unconjugated | |
KRT19 | |
40-kDa keratin intermediate filament; AI663979; C K1CS; CK 19; CK19; CK-19; cytokeratin 19; cytokeratin-19; EndoC; K19; K1CS; Ka19; keratin 19; keratin 19, type I; keratin complex 1, acidic, gene 19; keratin, type I cytoskeletal 19; keratin, type I, 40-kd; keratin-19; Krt-1.19; Krt1-19; Krt19; LOC100344434; MGC15366; Type I keratin Ka19 | |
Mouse | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG1 |