Learn More
Cytokeratin 18 Monoclonal Antibody (4D11E4), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA515458
Code : A0
Additional Details : Weight : 0.01000kg
Description
MA5-15458 targets Cytokeratin 18 in indirect ELISA, IHC and WB applications and shows reactivity with Human samples. The MA5-15458 immunogen is purified recombinant fragment of human Cytokeratin 18 (aa391-483) expressed in E. Coli.<br>. MA5-15458 detects Cytokeratin 18 which has a predicted molecular weight of approximately 48kDa.
Cytokeratin 18 Monoclonal antibody specifically detects Cytokeratin 18 in Human, Mouse samples. It is validated for ELISA, Immunohistochemistry, Western Blot.Specifications
Cytokeratin 18 | |
Monoclonal | |
Conc. Not Determined | |
ascites with 0.03% sodium azide | |
P05783, P05784 | |
KRT18 | |
Purified recombinant fragment of human Cytokeratin 18 (aa391-483) expressed in E. Coli. | |
RUO | |
16668, 3875 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
ELISA, Immunohistochemistry, Western Blot | |
4D11E4 | |
Unconjugated | |
KRT18 | |
cb83; Cell proliferation-inducing gene 46 protein; CK 18; CK18; CK-18; CYK18; cytokeratin 18; cytokeratin endo B; cytokeratin type -1; DAPK-1; Cytokeratin18; cytokeratin-18; dapk1; dorsal aorta proneprin kinesin-1; DreK18; Endo B; I79_023185; K 18; K18; keratin 18; keratin 18, type I; keratin 18a, tandem duplicate 1; keratin complex 1, acidic, gene 18; keratin D; Keratin, type I cytoskeletal 18; Keratin18; keratin-18; Kerd; KRT 18; Krt1-18; KRT18; krt18 {ECO:0000250; krt18a.1; LOC100008885; PIG46; sb:cb83; type I cytoskeletal 18; UniProtKB:P05783}; wu:fa13f02; wu:fb36b04 | |
Mouse | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG2b |