Learn More
CYP4B1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA528925
Code : PN
Additional Details : Weight : 0.01000kg
Description
Recommended positive controls: 293T, A431, H1299, HeLaS3, HepG2, Molt-4, Raji. Predicted reactivity: Mouse (85%), Rat (88%), Rabbit (87%), Bovine (82%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
CYP4B1 Polyclonal antibody specifically detects CYP4B1 in Human samples. It is validated for Western Blot.Specifications
CYP4B1 | |
Polyclonal | |
Unconjugated | |
CYP4B1 | |
CYP4B1; Cyp4b-1; CYP4B1 isoform; CYP4B1-like isozyme short form; CYPIVB1; cytochrome P450 4B1; cytochrome P450 family 4 subfamily B member 1; cytochrome P450 isozyme 5; cytochrome P-450 isozyme 5; Cytochrome P450 L-2; cytochrome P450, family 4, subfamily B, polypeptide 1; cytochrome P450, subfamily 4B, polypeptide 1; cytochrome P450, subfamily IV B, polypeptide 1; cytochrome P450, subfamily IVB, polypeptide 1; cytochrome P450-HP; microsomal monooxygenase; P450 L-2; P-450HP; P450-isozyme 5 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
1580 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot | |
0.69 mg/mL | |
0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7 | |
P13584 | |
CYP4B1 | |
Recombinant fragment corresponding to a region within amino acids 1 and 251 of Human Cytochrome P450 4B1 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |