missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CYP2D6 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA535148
Code : NEW
Additional Details : Weight : 0.01000kg
Description
This antibody is predicted to react with rat based on sequence homology.
CYP2D6 Polyclonal antibody specifically detects CYP2D6 in Human samples. It is validated for Immunohistochemistry (Paraffin), Western Blot.Specifications
CYP2D6 | |
Polyclonal | |
Unconjugated | |
CYP2D6 | |
AI303445; Cholesterol 25-hydroxylase; CPD6; CYP2D; Cyp2d10; Cyp2d-10; Cyp2d13; Cyp2d18; Cyp2d-18; Cyp2d22; Cyp2d3; Cyp2d-3; Cyp2d4; Cyp2d-4; Cyp2d4v1; Cyp2d4v2; Cyp2d6; CYP2D7AP; CYP2D7BP; CYP2D7P2; CYP2D8P2; CYP2DL1; CYPIID10; CYPIID18; CYPIID3; CYPIID4; CYPIID6; Cytochrome P450 2D10; cytochrome P450 2d18; Cytochrome P450 2D-29; Cytochrome P450 2D3; cytochrome P450 2D-35; cytochrome P450 2D4; Cytochrome P450 2D6; cytochrome P450 family 2 subfamily D member 6; cytochrome P450, 2d10; cytochrome P450, family 2, subfamily d, polypeptide 10; cytochrome P450, family 2, subfamily d, polypeptide 13; cytochrome P450, family 2, subfamily d, polypeptide 22; cytochrome P450, family 2, subfamily d, polypeptide 3; cytochrome P450, family 2, subfamily d, polypeptide 4; cytochrome P450, family 2, subfamily d, polypeptide 6; cytochrome P450, family 2, subfamily D, polypeptide 7 pseudogene 2; cytochrome P450, family 2, subfamily D, polypeptide 8 pseudogene 2; cytochrome P450, subfamily 2D, polypeptide | |
Rabbit | |
Antigen affinity chromatography, Protein A | |
RUO | |
1565 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.4 mg/mL | |
PBS with 0.09% sodium azide | |
P10635 | |
CYP2D6 | |
KLH conjugated synthetic peptide between 76-105 amino acids from the N-terminal region of human CYP2D6 | |
400 μL | |
Primary | |
Human | |
Antibody | |
IgG |