missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CYTL1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA538912
Code : NEW
Additional Details : Weight : 0.00900kg
Description
C17 is a cytokine-like protein specifically expressed in bone marrow and cord blood mononuclear cells that bear the CD34 (MIM 142230) surface marker (Liu et al., 2000 [PubMed 10857752]).[supplied by OMIM, Mar 2008].Specifications
CYTL1 | |
Polyclonal | |
Unconjugated | |
CYTL1 | |
C4orf4 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
54360 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
1 mg/mL | |
Dulbecco′s PBS with 150mM NaCl, 50% glycerol and 0.02% sodium azide; pH 7.4 | |
Q9NRR1 | |
CYTL1 | |
A synthetic peptide derived from the internal region of human CYTL1 | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG |