missing translation for 'onlineSavingsMsg'
Learn More

Cystatin/Stefin Homolog 2 Rat anti-Mouse, Clone: 237305, R&D Systems™

Rat Monoclonal Antibody

Brand:  RD Systems (Bio-Techne) MAB1516

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13488695

  • £353.77 / 500ยตg


Estimated Shipment
04-10-2018
Add to basket

Description

Description

Cystatin/Stefin Homolog 2 Monoclonal antibody MAB1516 Specifically detects Cystatin/Stefin Homolog 2 in Mouse samples. It is validated for Immunoprecipitation, Western Blotting
Specifications

Specifications

Cystatin/Stefin Homolog 2
237305
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Purified
Rat
IgG2b
500 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20 to -70 °C under sterile conditions after reconstitution.
Monoclonal
Detects mouse Cystatin/Stefin Homolog 2 in direct ELISAs and Western blots. In direct ELISAs and Western blots, no cross-reactivity with recombinant human (rh) Cystatin D, S, SA, SN, recombinant mouse Cystain B, C, E/M, rhFetuin A, rhFetuin B, rhHPRG, or rhKininostatin is observed.
Mouse
Western Blot, Immunoprecipitation
LYOPH
Western Blot 1 ug/mL, Immunoprecipitation 25 ug/mL
NP_084009
2010005H15Rik
E. coli-derived recombinant mouse Cystatin/Stefin Homolog 2, Met2-Phe97, Accession # NP_084009
Protein A or G purified from hybridoma culture supernatant
RUO
Primary
76770
Reconstitute at 0.5 mg/mL in sterile PBS.
Videos

For Research Use Only