missing translation for 'onlineSavingsMsg'
Learn More

Cystatin S Mouse anti-Human, Clone: 201805, R&D Systems™

Mouse Monoclonal Antibody

Brand:  RD Systems (Bio-Techne) MAB1296

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13458525

  • £337.93 / 500µg


Estimated Shipment
06-03-2019
Add to basket

Description

Description

Cystatin S Monoclonal antibody MAB1296 Specifically detects Cystatin S in Human samples. It is validated for Immunoprecipitation, Western Blotting
Specifications

Specifications

Cystatin S
201805
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Purified
CST4
Mouse myeloma cell line NS0-derived recombinant human Cystatin S Ser21-Ala141 Accession # P01036
Protein A or G purified from hybridoma culture supernatant
RUO
Primary
1472
Reconstitute at 0.5 mg/mL in sterile PBS.
Western Blot, Immunoprecipitation
LYOPH
Western Blot 1 ug/mL, Immunoprecipitation 25 ug/mL
P01036
CST4, cystatin 4, cystatin S, Cystatin-4, cystatin-S, cystatin-SA-III, MGC71923, Salivary acidic protein 1
Mouse
IgG2b
500 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Monoclonal
Detects human Cystatin S in direct ELISAs and Western blots. In direct ELISAs, 100% cross-reactivity with recombinant human (rh) Cystatin SA is observed and no cross-reactivity with rhCystatin D, E, SN, rhFetuin A, rhFetuin B, or rhHPRG (Histidine-Proline-Rich Glycoprotein) is observed. In Western blots, approximately 15% cross-reactivity with rhCystatins SA and SN (under reducing conditions only) is observed and no cross-reactivity with rhCystatin A, B, C, D, E/M, rhFetuin A, rhFetuin B, or rhHPRG is observed.
Human
Videos

For Research Use Only