Learn More
Cystatin F Mouse anti-Human, Clone: 292103, R&D Systems™
Mouse Monoclonal Antibody
Brand:  RD Systems (Bio-Techne) MAB1889
Code : Z1
Additional Details : Weight : 0.00010kg
 View more versions of this product
Product Code. 13439005
  • £313.76 / 100µg
Estimated Shipment
18-08-2020
Description
Description
Cystatin F Monoclonal antibody MAB1889 Specifically detects Cystatin F in Human samples. It is validated for Immunoprecipitation, Western Blotting
Specifications
Specifications
Cystatin F
292103
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Purified
CST7
Mouse myeloma cell line NS0-derived recombinant human Cystatin F Gly20-His145 Accession # O76096
Protein A or G purified from hybridoma culture supernatant
RUO
Primary
8530
Reconstitute at 0.5 mg/mL in sterile PBS.
Western Blot, Immunoprecipitation
LYOPH
Western Blot 1 ug/mL, Immunoprecipitation 25 ug/mL
O76096
CMAP, CST7, Cystatin 7, cystatin F (leukocystatin), Cystatin-7, cystatin-F, Cystatin-like metastasis-associated protein, leukocystatin
Mouse
IgG2b
100 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Monoclonal
Detects human Cystatin F in direct ELISAs and Western blots. In direct ELISAs and Western blots, no cross-reactivity with recombinant human (rh) Cystatin A, B, C, D, E/M, S, recombinant mouse (rm) Cystatin/Stefin Homolog, rhFetuin A, rhFetuin B, rhHPRG, rhKininogen, or rhKininostatin is observed.