missing translation for 'onlineSavingsMsg'
Learn More

Cystatin B Rat anti-Mouse, Clone: 227807, R&D Systems™

Rat Monoclonal Antibody

Brand:  RD Systems (Bio-Techne) MAB14091

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13438615

  • £376.32 / 500µg

Estimated Shipment date
from Supplier 19-11-2019

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Cystatin B Monoclonal antibody MAB14091 Specifically detects Cystatin B in Mouse samples. It is validated for Immunohistochemistry
Specifications

Specifications

Cystatin B/Stefin B
227807
Immunohistochemistry 8-25 ug/mL
Q62426
CPI-B, CST6cystatin B (liver thiol proteinase inhibitor)10STFBcystatin-B, CSTB, cystatin B (stefin B), EPM1, Liver thiol proteinase inhibitor, PME, stefin B, stefin-B
Rat
IgG2a
500 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Monoclonal
Detects mouse Cystatin B in direct ELISAs. In direct ELISAs, 100% cross-reactivity with recombinant human (rh) Cystatin B is observed and no cross-reactivity with rhCystatin A, C, D, E/M, F, S, SA, rhFetuin A, rhFetuin B, rhHPRG, or rhKininogen is observed.
Mouse
Immunohistochemistry
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Purified
CSTB
E. coli-derived recombinant mouse Cystatin B Met2-Phe98 Accession # Q62426
Protein A or G purified from hybridoma culture supernatant
RUO
Primary
1476
Reconstitute at 0.5 mg/mL in sterile PBS.
Videos
Documents

For Research Use Only