missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CYSLTR2 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA533465
Code : NEW
Additional Details : Weight : 0.01000kg
Description
Percent identity with other species by BLAST analysis: Human, Gorilla (100%); Horse (80%). Immunohistochemistry with formalin-fixed paraffin-embedded tissues requires pretreatment with Proteinase K.
CYSLTR2 Polyclonal antibody specifically detects CYSLTR2 in Human samples. It is validated for Immunohistochemistry (Paraffin).Specifications
CYSLTR2 | |
Polyclonal | |
Unconjugated | |
CYSLTR2 | |
2300001H05Rik; Cltr2; CysLT(2); Cyslt2; CysLT-2; CYSLT2R; CYSLTR2; cysteinyl leukotriene CysLT2 receptor; cysteinyl leukotriene receptor 2; GPCR21; G-protein coupled receptor GPCR21; G-protein coupled receptor HG57; HG57; hGPCR21; HPN321; KPG_011; PSEC0146; RSBPT32 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
57105 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
1 mg/mL | |
PBS with 0.1% sodium azide | |
Q9NS75 | |
CYSLTR2 | |
Synthetic 15 amino acid peptide from N-Terminus of human CYSLTR2 / CYSLT2. | |
50 μg | |
Primary | |
Human | |
Antibody | |
IgG |