missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CYR61 Polyclonal Antibody, DyLight™ 650, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA522763
Code : LD
Additional Details : Weight : 0.01000kg
Description
This antibody is predicted to react with rat and mouse based on 88% sequence homology.
CYR61 Polyclonal antibody specifically detects CYR61 in Human, Mouse, Rabbit samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot.Specifications
CYR61 | |
Polyclonal | |
DyLight 650 | |
CCN1 | |
3CH61; AI325051; CCN family member 1; CCN1; cellular communication network factor 1; Cyr61; cysteine rich angiogenic inducer 61; cysteine rich protein 61; cysteine-rich angiogenic inducer 61; cysteine-rich heparin-binding protein 61; cysteine-rich, angiogenic inducer, 61; cysteine-rich, anigogenic inducer, 61; GIG1; IBP-10; IGF-binding protein 10; IGFBP10; IGFBP-10; insulin-like growth factor binding protein 10; insulin-like growth factor-binding protein 10; intermediate early; protein CYR61; Protein GIG1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
100339657, 16007, 3491 | |
4° C, store in dark | |
Liquid |
Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot | |
1 mg/mL | |
50mM sodium borate with no preservative | |
O00622, P18406 | |
CCN1 | |
A synthetic peptide made to the human CYR61 protein sequence (between residues 150-250). | |
100 μL | |
Primary | |
Human, Mouse, Rabbit | |
Antibody | |
IgG |