Learn More
Cyr61/CCN1 Mouse anti-Human, Clone: 365108, R&D Systems™
Mouse Monoclonal Antibody
Brand:  RD Systems (Bio-Techne) MAB4055
Code : Z1
Additional Details : Weight : 0.00010kg
 View more versions of this product
Product Code. 13430386
  • £311.64 / 100µg
Estimated Shipment date
from Supplier 30-03-2020
Description
Description
Cyr61/CCN1 Monoclonal antibody MAB4055 Specifically detects Cyr61/CCN1 in Human samples. It is validated for Western Blotting
Specifications
Specifications
Cyr61/CCN1
365108
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Purified
CYR61
Mouse myeloma cell line NS0-derived recombinant human Cyr61 Ala22-Asp381 Accession # O00622
Protein A or G purified from hybridoma culture supernatant
RUO
Primary
3491
Reconstitute at 0.5 mg/mL in sterile PBS.
Western Blot
LYOPH
Western Blot 1 ug/mL
O00622
CCN family member 1, CCN1, CCN1IGF-binding protein 10, Cysteine-rich angiogenic inducer 61, cysteine-rich, angiogenic inducer, 61, GIG1, GIG1cysteine-rich heparin-binding protein 61, IBP-10, IGFBP-10, IGFBP10cysteine-rich, anigogenic inducer, 61, Insulin-like growth factor-binding protein 10, protein CYR61, Protein GIG1
Mouse
IgG1
100 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Monoclonal
Detects human Cyr61/CCN1 in direct ELISAs and Western blots.