missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cyclosporin A Monoclonal Antibody (CSZ.22), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA191110
Code : LE
Additional Details : Weight : 0.01000kg
Description
MA1-91110 detects Cyclosporin A from various samples.
Cyclosporin A Monoclonal antibody specifically detects Cyclosporin A in Chemical samples. It is validated for ELISA.Specifications
Cyclosporin A | |
Monoclonal | |
5 mg/mL | |
PBS with 0.09% sodium azide; pH 7.4 | |
Mouse | |
Protein G | |
RUO | |
4° C, do not freeze | |
Liquid | |
Chemical |
ELISA | |
CSZ.22 | |
Unconjugated | |
C62H111N11O12; Ciclosporin; Cyclophilin A; Cyclosporin; Cyclosporin A-binding protein; Cyclosporine; Cyclosporine A; CYPA; CYPH; epididymis secretory sperm binding protein Li 69p; HEL-S-69p; Peptidyl-prolyl cis-trans isomerase A; Peptidyl-prolyl cis-trans isomerase A, N-terminally processed; peptidylprolyl isomerase A; peptidylprolyl isomerase A (cyclophilin A); PPIA; PPIase A; rotamase A; T cell cyclophilin | |
Proprietary immunogen | |
100 μg | |
Primary | |
Antibody | |
IgG1 |