missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cyclin E Monoclonal Antibody (HE172), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen 321500
Code : 50
Additional Details : Weight : 0.02000kg
Description
Cyclin E Monoclonal antibody specifically detects Cyclin E in Human samples. It is validated for Gel Shift, Immunocytochemistry, Immunoprecipitation, Western Blot.Specifications
Cyclin E | |
Monoclonal | |
0.5 mg/mL | |
PBS with 0.1% sodium azide; pH 7.4 | |
P24864 | |
CCNE1 | |
This antibody was prepared against full-length, recombinant, human cyclin E protein. | |
200 μg | |
Primary | |
Human | |
Antibody | |
IgG2b κ |
Gel Shift, Immunocytochemistry, Immunoprecipitation, Western Blot | |
HE172 | |
Unconjugated | |
CCNE1 | |
AW538188; Ccne; Ccne1; CycE1; CYCLE; cyclin E; cyclin E1; cyclin Es; cyclin Et; E-S; G1/S-specific cyclin-E1; pCCNE1 | |
Mouse | |
Protein A | |
RUO | |
898 | |
-20°C | |
Liquid |