Learn More
Cyclin D3 Mouse, Unlabeled, Clone: 1, BD
Mouse Monoclonal Antibody
Brand: BD Biosciences 610279
Additional Details : Weight : 0.01100kg
Description
Several classes of cyclins (A-E) have been described. These proteins act as regulatory subunits for cyclin-dependent kinases (cdks). The synthesis and degradation of cyclins is tightly controlled in a cell cycle specific manner. There are at least 3 different D-type cyclins whose relative levels vary among cell types. Cyclin D3 is known to associate with Cdk5 and weakly with Cdk2. Cdk4 is preferentially bound by Cyclin D1. However, in T cells which do not express Cyclin D1, Cdk4 associates with Cyclins D2 and D3. While bound to cdks, the D-type cyclins also associate with the polymerase-δ subunit, PCNA. Cyclin D3 has also been shown to bind pRb in vitro. This suggests that pRb may be an in vivo substrate of D-type cyclins.
This antibody is routinely tested by western blot analysis. Other applications were tested at BD Biosciences Pharmingen during antibody development only or reported in the literature.
Specifications
Cyclin D3 | |
Monoclonal | |
250μg/mL | |
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide. | |
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide. | |
50 μg | |
Cell Biology | |
Canine, Human, Mouse, Rat | |
IgG2b |
Western Blot | |
1 | |
Unconjugated | |
Mouse | |
Affinity Purified | |
RUO | |
Primary | |
Store undiluted at -20°C. |
For Research Use Only.