Learn More
CXCR4 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA3305
Code : LE
Additional Details : Weight : 0.01000kg
Description
Several different molecular weight isoforms of CXCR4 have been described in the literature, which vary depending on cell type tested and extraction buffer used. Using Ramos, Raji, and Jurkat membrane fractions, two different MW forms of CXCR4 were detected with PA3-305 by Western blot, between 55-72kD. It is recommended to use a membrane isolation buffer/kit for extracting CXCR4 for Western blot analysis. Cellular membrane fractions were isolated for Western blot using Mem-PER Plus (Product # 89842).
CXCR4 Polyclonal antibody specifically detects CXCR4 in Human, Mouse samples. It is validated for Flow Cytometry, Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot.Specifications
CXCR4 | |
Polyclonal | |
Unconjugated | |
Cxcr4 | |
b2b220Clo; C Cmotif chemokine; C X C motif chemokine; CC motif chemokine; CCmotif chemokine; CD antigen CD184; CD184; CD184 antigen; chemokine (C-X-C motif) receptor 4; chemokine (C-X-C) receptor 4; chemokine receptor; chemokine receptor (LCR1); chemokine receptor 4; chemokine receptor CXCR4; Cmkar4; CXC; CXC chemokine receptor; C-X-C chemokine receptor type 4; CXC motif chemokine; C-X-C motif chemokine receptor 4; CXCR4; CXC-R4; CXCR-4; D2S201E; FB22; Fusin; HM89; HSY3RR; HUMSTR; LAP3; LAP-3; LCR1; LESTR; Leukocyte-derived seven transmembrane domain receptor; leukocyte-expressed seven-transmembrane-domain; lipopolysaccharide-associated protein 3; LPS-associated protein 3; neuropeptide Y receptor Y3; neuropeptide Y3 receptor; NPY3R; NPYR; NPYRL; NPYY3R; PB-CKR; PBSF/SDF-1; pre-B-cell-derived chemokine receptor; SDF-1 receptor; Sdf1r; seven transmembrane helix receptor; seven-transmembrane-segment receptor, spleen; stromal cell-derived factor 1 receptor; WHIM; WHIMS | |
Rabbit | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Flow Cytometry, Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot | |
Conc. Not Determined | |
whole serum with 0.05% sodium azide | |
P61073, P70658 | |
Cxcr4 | |
Synthetic peptide corresponding to residues K(338) G K R G G H S S V S T E S E S S S F H S S (359) of human CXCR4 receptor protein. | |
RUO | |
12767, 7852 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |