Learn More
CXCL7/Thymus Chemokine-1 Rat anti-Mouse, Clone: 159703, R&D Systems™
Rat Monoclonal Antibody
Brand:  RD Systems (Bio-Techne) MAB10911
Code : Z1
Additional Details : Weight : 0.00010kg
 View more versions of this product
Product Code. 13448325
  • £388.08 / 500µg
Estimated Shipment date
from Supplier 30-03-2020
Description
Description
CXCL7/Thymus Chemokine-1 Monoclonal antibody MAB10911 Specifically detects CXCL7/Thymus Chemokine-1 in Mouse samples. It is validated for ELISA Capture, Western Blotting
Specifications
Specifications
CXCL7/Thymus Chemokine-1
159703
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Purified
Rat
IgG2a
500 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Monoclonal
Detects mouse CXCL7/Thymus Chemokine-1 in ELISAs and Western blots.
Mouse
Western Blot, ELISA
LYOPH
Western Blot 1 ug/mL, ELISA Capture (Matched Antibody Pair) 2-8 ug/mL
Q9EQI5
Ppbp
E. coli-derived recombinant mouse CXCL7/Thymus Chemokine-1 Lys40-Tyr113 Accession # Q9EQI5
Protein A or G purified from hybridoma culture supernatant
RUO
Primary
5473
Reconstitute at 0.5 mg/mL in sterile PBS.