missing translation for 'onlineSavingsMsg'
Learn More

CXCL14/BRAK Mouse anti-Human, Clone: 131120, R&D Systems™

Mouse Monoclonal Antibody

Brand:  RD Systems (Bio-Techne) MAB866

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13461936

  • £337.93 / 500µg


Estimated Shipment
04-03-2019
Add to basket

Description

Description

CXCL14/BRAK Monoclonal antibody MAB866 Specifically detects CXCL14/BRAK in Human samples. It is validated for ELISA Capture, Western Blotting
Specifications

Specifications

CXCL14/BRAK
131120
Western Blot 1 ug/mL, ELISA Capture (Matched Antibody Pair) 2-8 ug/mL
O95715
BMAC, Bolekine, BRAK, BRAKKEC, chemokine (C-X-C motif) ligand 14, Chemokine BRAK, CXC-X3, member 14 (BRAK), MIP-2 gamma, MIP2G, MIP-2G, NJACKec, SCYB14MGC10687, Small-inducible cytokine B14
Mouse
IgG2a
500 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Monoclonal
Detects human CXCL14/BRAK in ELISAs and Western blots. In direct ELISAs, shows approximately 80% cross-reactivity with recombinant mouse (rm) CXCL14/BRAK and no cross-reactivity with recombinant human (rh)GRO alpha , rhGRO beta , rhGRO gamma ,rhMIP-1 alpha , rmMIP-1 alpha , rmMIP-1 beta , rhMIP-1 delta , rmMIP-1 gamma , rmMIP-2, rhMIP-3 alpha , rmMIP-3 alpha , rrMIP-3 alpha , rhMIP-3 beta , or rmMIP-3 beta .
Human
Western Blot, ELISA
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Purified
CXCL14
E. coli-derived recombinant human CXCL14/BRAK Leu23-Tyr99 Accession # O95715
Protein A or G purified from hybridoma culture supernatant
RUO
Primary
9547
Reconstitute at 0.5 mg/mL in sterile PBS.
Videos

For Research Use Only