missing translation for 'onlineSavingsMsg'
Learn More

CXCL1/2/3/GRO Pan Specific Mouse anti-Human, Clone: 29702, R&D Systems™

Mouse Monoclonal Antibody

Brand:  RD Systems (Bio-Techne) MAB2761

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13459675

  • £249.49 / 100µg


Estimated Shipment
28-02-2019
Add to basket

Description

Description

CXCL1/2/3/GRO Pan Specific Monoclonal antibody MAB2761 Specifically detects CXCL1/2/3/GRO Pan Specific in Human samples. It is validated for Flow Cytometry (Intracellular Staining), Western Blotting
Specifications

Specifications

CXCL1/2/3/GRO
29702
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Purified
E. coli-derived recombinant human CXCL3 Ala35-Asn107 Accession # P19876
Protein A or G purified from hybridoma culture supernatant
RUO
Primary
2919
Reconstitute at 0.5 mg/mL in sterile PBS.
Western Blot, Flow Cytometry, CyTOF
LYOPH
Western Blot 1 ug/mL, Intracellular Staining by Flow Cytometry 2.5 ug/10^6 cells, CyTOF-ready
P19876
Mouse
IgG1
100 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Monoclonal
Detects human CXCL1/2/3 in direct ELISAs and Western blots. In direct ELISAs, 100% cross-reactivity with recombinant mouse (rm) CXCL2 is observed and no cross-reactivity with recombinant human (rh) VIC, rmVIC, rhCCL3L1, rhCCL4L1, or recombinant canine IL-8 is observed.
Human
Videos

For Research Use Only