Learn More
Cryptic Mouse anti-Human, Clone: 219708, R&D Systems™
Mouse Monoclonal Antibody
Brand:  RD Systems (Bio-Techne) MAB1410
Code : Z1
Additional Details : Weight : 0.00010kg
 View more versions of this product
Product Code. 13458615
  • £337.93 / 500µg
Estimated Shipment
06-03-2019
Add to basket
Description
Description
Cryptic Monoclonal antibody MAB1410 Specifically detects Cryptic in Human samples. It is validated for Western Blotting
Specifications
Specifications
CYRPTIC/CFC1
219708
Western Blot 1 ug/mL
P0CG36
CFC1, CFC-1, CFC1B, cripto, FRL-1, cryptic family 1, Cryptic family protein 1, cryptic protein, CRYPTICFLJ77897, HTX2MGC133213
Mouse
IgG1
500 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Monoclonal
Detects human Cryptic in direct ELISAs and Western blots. In direct ELISAs and Western blots, no cross-reactivity with recombinant mouse Cryptic is observed.
Human
Western Blot
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Purified
CFC1
Mouse myeloma cell line NS0-derived recombinant human Cryptic Tyr26-Gly169 Accession # P0CG36
Protein A or G purified from hybridoma culture supernatant
RUO
Primary
55997
Reconstitute at 0.5 mg/mL in sterile PBS.