Learn More
CRTH-2 Mouse anti-Human, Clone: 301108, R&D Systems™
Mouse Monoclonal Antibody
Brand:  RD Systems (Bio-Techne) MAB3338
Code : Z1
Additional Details : Weight : 0.00010kg
 View more versions of this product
Product Code. 13450016
  • £311.64 / 100µg
Estimated Shipment date
from Supplier 20-11-2019
Description
Description
CRTH-2 Monoclonal antibody MAB3338 Specifically detects CRTH-2 in Human samples. It is validated for Flow Cytometry
Specifications
Specifications
CRTH-2/GPR44
301108
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS and NaCl with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Purified
PTGDR2
NS0 mouse myeloma cell line transfected with human CRTH-2 Met1-Ser395 Accession # Q9Y5Y4
Protein A or G purified from hybridoma culture supernatant
RUO
Primary
11251
Reconstitute at 0.5 mg/mL in sterile PBS.
Flow Cytometry, CyTOF
LYOPH
Flow Cytometry 2.5 ug/10^6 cells, CyTOF-ready
Q9Y5Y4
CD294, CD294 antigen, Chemoattractant receptor-homologous molecule expressed on Th2 cells, CRTH2, CRTH2chemoattractant receptor homologous molecule expressed on T helper type 2 cells, DL1R, DP2, G protein-coupled receptor 44, GPR44, PGD2R, PTGDR2, putative G-protein coupled receptor 44
Mouse
IgG2a
100 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Monoclonal
Detects human CRTH-2. Stains human CRTH-2-transfected cells but not parental cells.