missing translation for 'onlineSavingsMsg'
Learn More

Crossveinless-2/CV-2 Rat anti-Mouse, Clone: 349920, R&D Systems™

Rat Monoclonal Antibody

Brand:  RD Systems (Bio-Techne) MAB2299

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13449335

  • £277.83 / 100µg

Call for latest availability
+44 (0)1509 555 500

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Crossveinless-2/CV-2 Monoclonal antibody MAB2299 Specifically detects Crossveinless-2/CV-2 in Mouse samples. It is validated for Western Blotting
Specifications

Specifications

Crossveinless-2/CV-2/BMPER
349920
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Purified
BMPER
Mouse myeloma cell line NS0-derived recombinant mouse Crossveinless-2/CV-2 Val34-Arg685 Accession # AAH66153
Protein A or G purified from hybridoma culture supernatant
RUO
Primary
168667
Reconstitute at 0.5 mg/mL in sterile PBS.
Western Blot
LYOPH
Western Blot 1 ug/mL
AAH66153
BMP binding endothelial regulator, BMP-binding endothelial regulator precursor protein, BMP-binding endothelial regulator protein, BMPER, Bone morphogenetic protein-binding endothelial cell precursor-derived regulator, CRIM3, crossveinless 2, crossveinless-2, Cv2, CV-2, hCV2, KIAA1965, Protein crossveinless-2
Rat
IgG2a
100 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Monoclonal
Detects mouse Crossveinless-2/CV-2 in direct ELISAs and Western blots. In direct ELISAs and Western blots, 100% cross-reactivity with recombinant human CV-2 is observed.
Mouse
Videos
Documents

For Research Use Only