Learn More
CRIM1 Mouse anti-Human, Clone: 253511, R&D Systems™
Mouse Monoclonal Antibody
Brand:  RD Systems (Bio-Techne) MAB1917
Code : Z1
Additional Details : Weight : 0.00010kg
 View more versions of this product
Product Code. 13449025
  • £376.32 / 500µg
Estimated Shipment date
from Supplier 20-11-2019
Description
Description
CRIM1 Monoclonal antibody MAB1917 Specifically detects CRIM1 in Human samples. It is validated for Western Blotting
Specifications
Specifications
CRIM1
253511
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Purified
CRIM1
Mouse myeloma cell line NS0-derived recombinant human CRIM1 Leu35-Ser939 Accession # Q9NZV1
Protein A or G purified from hybridoma culture supernatant
RUO
Primary
51232
Reconstitute at 0.5 mg/mL in sterile PBS.
Western Blot
LYOPH
Western Blot 1 ug/mL
Q9NZV1
CRIM-1, cysteine rich transmembrane BMP regulator 1 (chordin-like), cysteine-rich motor neuron 1, cysteine-rich motor neuron 1 protein, Cysteine-rich repeat-containing protein S52, MGC138194, S52
Mouse
IgG1
500 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Monoclonal
Detects human CRIM1 in direct ELISAs and Western blots.