missing translation for 'onlineSavingsMsg'
Learn More

CRHR1 Mouse anti-Human, Clone: 343919, R&D Systems™

Mouse Monoclonal Antibody

Brand:  RD Systems (Bio-Techne) MAB3930

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13460326

  • £271.93 / 100µg


Estimated Shipment
21-12-2018
Add to basket

Description

Description

CRHR1 Monoclonal antibody MAB3930 Specifically detects CRHR1 in Human samples. It is validated for Flow Cytometry, Immunohistochemistry
Specifications

Specifications

CRHR1/CRF1
343919
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Purified
CRHR1
NS0 mouse myeloma cell line transfected with human CRHR-1, Ser24-Val415, Accession # NP_004373
Protein A or G purified from hybridoma culture supernatant
RUO
Primary
1394
Reconstitute at 0.5 mg/mL in sterile PBS.
Flow Cytometry, Immunohistochemistry, CyTOF
LYOPH
Flow Cytometry 2.5 ug/10^6 cells, Immunohistochemistry 8-25 ug/mL, CyTOF-ready
NP_004373
CHR-R1, corticotropin releasing hormone receptor 1, corticotropin-releasing factor receptor 1, corticotropin-releasing factor type 1 receptor, Corticotropin-releasing hormone receptor 1, CRF1, CRFR, CRF-R, CRFR1, CRF-R1, CRF-R-1, CRFR-1, CRH-R-1, CRHR1f, CRH-R1h, CRHR1L, CRHRCRH-R1, seven transmembrane helix receptor
Mouse
IgG2a
100 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20 to -70 °C under sterile conditions after reconstitution.
Monoclonal
Detects human CRHR-1 transfectants but not irrelevant transfectants.
Human
Videos

For Research Use Only