missing translation for 'onlineSavingsMsg'
Learn More
Learn More
COX2 Monoclonal Antibody (SP21), Invitrogen™
Rabbit Monoclonal Antibody
Brand: Invitrogen MA514568
Code : A0
Additional Details : Weight : 0.01000kg
Description
MA5-14568 targets COX2 in IHC (P) applications and shows reactivity with Human samples. The MA5-14568 immunogen is a synthetic peptide from C-terminus of rat cox2.
COX2 Monoclonal antibody specifically detects COX2 in Human samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot.Specifications
COX2 | |
Monoclonal | |
0.006 mg/mL | |
PBS with 1% BSA and 0.1% sodium azide; pH 7.2 | |
P35354 | |
PTGS2 | |
A synthetic peptide from C-terminus of rat cox2 | |
RUO | |
5743 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot | |
SP21 | |
Unconjugated | |
PTGS2 | |
COII; COX2; Cox-2; COXII; cyclooxygenase; cyclooxygenase 2; cyclooxygenase 2b; cyclooxygenase-2; Cytochrome c oxidase polypeptide II; Cytochrome c oxidase subunit 2; cytochrome c oxidase subunit II; glucocorticoid-regulated inflammatory cyclooxygenase; Gripghs; hCox-2; Macrophage activation-associated marker protein P71/73; mitochondrially encoded cytochrome c oxidase II; MTCO2; MT-CO2; PES-2; PGG/HS; PGH synthase 2; Pghs2; PGHS-2; Pghs-b; PHS II; PHS-2; prostaglandin G/H synthase 2; Prostaglandin H2 synthase 2; prostaglandin-endoperoxide synthase 2; prostaglandin-endoperoxide synthase 2 (prostaglandin G/H synthase and cyclooxygenase); Ptgs2; Tis10; TIS10 protein | |
Rabbit | |
500 μL | |
Primary | |
Human | |
Antibody | |
IgG |