missing translation for 'onlineSavingsMsg'
Learn More

COPD, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Brand:  Invitrogen OSC00035W

Code : LD

Additional Details : Weight : 0.01000kg

Product Code. 11540463

  • £257.00 / 100┬ÁL


Estimated Shipment
08-06-2018
Add to basket

Description

Description

Reconstitute with 100 ul of distilled water.
Specifications

Specifications

delta COP
Polyclonal
whole serum with no preservative
P48444, Q5XJY5, Q66H80
Archain, ARCN1, Coatomer subunit delta, Delta-coat protein, Delta-COP
Rabbit
100μL
Antibody
Polyclonal
Rat
Western Blot
Unconjugated
ARCN1
Serum
ARCN1
A synthetic peptide from human COPD
avoiding freeze/thaw cycles., store at -20°C, store at 4°C short term. For long term storage
Primary
372
Videos