missing translation for 'onlineSavingsMsg'
Learn More

COPD, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Brand:  Invitrogen OSC00036G

Code : LD

Additional Details : Weight : 0.01000kg

Product Code. 11550463

  • £257.00 / 500µg


Estimated Shipment
11-05-2018
Add to basket

Description

Description

Reconstitute with 500 ul of distilled water.
Specifications

Specifications

delta COP
Polyclonal
Unconjugated
ARCN1
Purified
ARCN1
A synthetic peptide from the internal region of human COPD conjugated to an immunogenic carrier protein was used as the antigen
Ammonium sulfate precipitation
avoiding freeze/thaw cycles., store at -20°C, store at 4°C short term. For long term storage
Primary
372
Western Blot
1mg/mL
PBS with no preservative
P48444, Q5XJY5, Q66H80
Archain, ARCN1, Coatomer subunit delta, Delta-coat protein, Delta-COP
Rabbit
IgG
500μg
Antibody
Polyclonal
Rat
Videos