missing translation for 'onlineSavingsMsg'
Learn More

Contactin 5, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Brand:  Invitrogen OSC00015W

Code : LD

Additional Details : Weight : 0.01000kg

Product Code. 11500453

  • £257.00 / 100┬ÁL


Estimated Shipment
16-05-2018
Add to basket

Description

Description

Reconstitute with 100 ul of distilled water.
Specifications

Specifications

Contactin 5
Polyclonal
whole serum with no preservative
O94779
Contactin 5, Neural recognition molecule NB-2, NB-2, hNB-2, HNB-2s, CNTN5, MGC163491
Rabbit
100μL
Antibody
Polyclonal
Human
Western Blot
Unconjugated
CNTN5
Serum
CNTN5
A synthetic peptide from human contactin-5
avoiding freeze/thaw cycles., store at -20°C, store at 4°C short term. For long term storage
Primary
53942
Videos