missing translation for 'onlineSavingsMsg'
Learn More

Contactin 5, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Brand:  Invitrogen OSC00021W

Code : LD

Additional Details : Weight : 0.01000kg

Product Code. 11560453

  • £257.00 / 100ĀµL


Estimated Shipment
06-07-2018
Add to basket

Description

Description

Reconstitute with 100 ul of distilled water.
Specifications

Specifications

Contactin 5
Polyclonal
whole serum with no preservative
P97527
NB-2, contactin 5, CNTN5, Neural recognition molecule NB-2
Rabbit
100μL
Antibody
Polyclonal
Rat
ImmunohistochemistryĆ¾Western Blot
Unconjugated
Cntn5
Serum
CNTN5
A synthetic peptide from rat Contactin-5
store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
114589
Videos